Assay Details
| Gene Symbol: cpna-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381505.2 | NP_001367323.1 | 6 - 7 | 1212 | 81 | |
| NM_063221.5 | NP_495622.2 | 6 - 7 | 1216 | 81 | ||
| NM_182097.5 | NP_871897.2 | 5 - 6 | 1114 | 81 | ||
More Information
Set Membership: |
|

cpna-5
Genomic Map