Assay Details
| Gene Symbol: din-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381614.1 | NP_001367110.1 | 1 - 2 | 125 | 67 | |
| NM_064083.6 | NP_496484.3 | 1 - 2 | 123 | 67 | ||
| NM_064084.8 | NP_496485.2 | 1 - 2 | 125 | 67 | ||
| NM_182101.5 | NP_871901.1 | 1 - 2 | 100 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381614.1 transcripts.
Set Membership: |
|

din-1
Genomic Map