Assay Details
| Gene Symbol: ubc-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361850.2 | NP_001348718.1 | 1 - 2 | 83 | 78 | |
| NM_001383804.1 | 2 - 3 | 92 | 78 | |||
| Gene Symbol: sqv-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061993.8 | NP_494394.1 | 1 - 2 | 85 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_061993.8 transcripts.
Set Membership: |
|

ubc-26
Genomic Map