Assay Details
| Gene Symbol: mak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373775.3 | NP_001359655.1 | 2 - 3 | 276 | 75 | |
| NM_001373776.2 | NP_001359681.1 | 3 | 281 | 75 | ||
| NM_063459.5 | NP_495860.1 | 2 - 3 | 270 | 75 | ||
More Information
Set Membership: |
|

mak-1
Genomic Map