Assay Details
| Gene Symbol: mak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027067.5 | NP_001022238.1 | 1 - 2 | 243 | 76 | |
| NM_001373775.3 | NP_001359655.1 | 3 - 4 | 775 | 76 | ||
| NM_001373776.2 | NP_001359681.1 | 3 - 4 | 780 | 76 | ||
| NM_001381538.1 | NP_001367107.1 | 1 - 2 | 243 | 76 | ||
| NM_063459.5 | NP_495860.1 | 3 - 4 | 769 | 76 | ||
More Information
Set Membership: |
|

mak-1
Genomic Map