Assay Details
| Gene Symbol: aqp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381493.1 | NP_001367603.1 | 4 - 5 | 442 | 77 | |
| NM_063109.6 | NP_495510.1 | 4 - 5 | 425 | 77 | ||
More Information
Set Membership: |
|

aqp-1
Genomic Map