Assay Details
| Gene Symbol: aqp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381493.1 | NP_001367603.1 | 5 - 6 | 765 | 81 | |
| NM_063109.6 | NP_495510.1 | 5 - 6 | 748 | 81 | ||
More Information
Set Membership: |
|

aqp-1
Genomic Map