Assay Details
| Gene Symbol: alh-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027041.8 | NP_001022212.1 | 3 - 4 | 366 | 94 | |
| NM_061545.7 | NP_493946.1 | 3 - 4 | 365 | 94 | ||
| NM_182151.6 | NP_871951.1 | 3 - 4 | 364 | 94 | ||
More Information
Set Membership: |
|

alh-6
Genomic Map