Assay Details
| Gene Symbol: alh-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047317.6 | NP_001040782.1 | 2 - 3 | 718 | 67 | |
| NM_061545.7 | NP_493946.1 | 5 - 6 | 1141 | 67 | ||
| NM_182151.6 | NP_871951.1 | 5 - 6 | 1140 | 67 | ||
More Information
Set Membership: |
|

alh-6
Genomic Map