Assay Details
| Gene Symbol: cpna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062335.7 | NP_494736.1 | 5 - 6 | 1355 | 64 | |
| NM_062336.5 | NP_494737.1 | 5 - 6 | 1355 | 64 | ||
More Information
Set Membership: |
|

cpna-1
Genomic Map