Assay Details
| Gene Symbol: alg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061436.5 | NP_493837.1 | 4 - 5 | 1679 | 67 | |
| NM_182192.5 | NP_871992.1 | 5 - 6 | 1774 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alg-2| Gene Symbol: alg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061436.5 | NP_493837.1 | 4 - 5 | 1679 | 67 | |
| NM_182192.5 | NP_871992.1 | 5 - 6 | 1774 | 67 | ||
Set Membership: |
|