Assay Details
| Gene Symbol: ctns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063303.6 | NP_495704.1 | 3 - 4 | 575 | 97 | |
| NM_182222.6 | NP_872022.1 | 3 - 4 | 577 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ctns-1| Gene Symbol: ctns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063303.6 | NP_495704.1 | 3 - 4 | 575 | 97 | |
| NM_182222.6 | NP_872022.1 | 3 - 4 | 577 | 97 | ||
Set Membership: |
|