Assay Details
| Gene Symbol: plpp-1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001355092.3 | NP_001342020.1 | 2 | 153 | 63 | |
| NM_001355093.2 | NP_001342021.1 | 1 - 2 | 142 | 63 | ||
| NM_001355094.2 | NP_001342019.1 | 1 - 2 | 142 | 63 | ||
| NM_062295.5 | NP_494696.1 | 1 - 2 | 142 | 63 | ||
| NM_182225.6 | NP_872025.1 | 1 - 2 | 151 | 63 | ||
| NM_182226.5 | NP_872026.1 | 1 - 2 | 148 | 63 | ||
More Information
Set Membership: |
|

plpp-1.1
Genomic Map