Assay Details
| Gene Symbol: plpp-1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182226.5 | NP_872026.1 | 4 - 5 | 1031 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plpp-1.1| Gene Symbol: plpp-1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182226.5 | NP_872026.1 | 4 - 5 | 1031 | 100 | |
Set Membership: |
|