Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267480.3 | NP_001254409.1 | 1 - 2 | 386 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cup-14| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267480.3 | NP_001254409.1 | 1 - 2 | 386 | 60 | |
Set Membership: |
|