Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267478.2 | NP_001254407.1 | 9 - 10 | 1814 | 80 | |
| NM_001267479.3 | NP_001254408.1 | 8 - 9 | 1784 | 80 | ||
| NM_001267480.3 | NP_001254409.1 | 2 - 3 | 468 | 80 | ||
| NM_001267481.4 | NP_001254410.1 | 2 - 3 | 244 | 80 | ||
More Information
Set Membership: |
|

cup-14
Genomic Map