Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267478.2 | NP_001254407.1 | 10 - 11 | 2200 | 84 | |
| NM_001267479.3 | NP_001254408.1 | 9 - 10 | 2170 | 84 | ||
| NM_001267480.3 | NP_001254409.1 | 3 - 4 | 854 | 84 | ||
| NM_001267481.4 | NP_001254410.1 | 3 - 4 | 630 | 84 | ||
| NM_001267482.3 | NP_001254411.1 | 1 - 2 | 352 | 84 | ||
More Information
Set Membership: |
|

cup-14
Genomic Map