Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267478.2 | NP_001254407.1 | 11 - 12 | 2725 | 80 | |
| NM_001267479.3 | NP_001254408.1 | 10 - 11 | 2695 | 80 | ||
| NM_001267480.3 | NP_001254409.1 | 4 - 5 | 1379 | 80 | ||
| NM_001267481.4 | NP_001254410.1 | 4 - 5 | 1155 | 80 | ||
| NM_001267482.3 | NP_001254411.1 | 2 - 3 | 877 | 80 | ||
More Information
Set Membership: |
|

cup-14
Genomic Map