Assay Details
| Gene Symbol: sra-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182260.4 | NP_872060.2 | 1 - 2 | 129 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sra-34| Gene Symbol: sra-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182260.4 | NP_872060.2 | 1 - 2 | 129 | 140 | |
Set Membership: |
|