Assay Details
| Gene Symbol: wnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268439.4 | NP_001255368.1 | 14 - 15 | 5524 | 61 | |
| NM_001268440.3 | NP_001255369.1 | 12 - 13 | 4720 | 61 | ||
| NM_001268441.3 | NP_001255370.1 | 14 - 15 | 5523 | 61 | ||
| NM_001268442.4 | NP_001255371.1 | 12 - 13 | 4711 | 61 | ||
| NM_001268443.3 | NP_001255372.1 | 13 - 14 | 5197 | 61 | ||
| NM_001268444.3 | NP_001255373.1 | 12 - 13 | 4714 | 61 | ||
| NM_001268445.3 | NP_001255374.1 | 11 - 12 | 5044 | 61 | ||
| NM_001268446.3 | NP_001255375.1 | 11 - 12 | 5035 | 61 | ||
| NM_001268447.3 | NP_001255376.1 | 11 - 12 | 5038 | 61 | ||
| NM_001268448.3 | NP_001255377.1 | 11 - 12 | 5029 | 61 | ||
| NM_001307110.3 | NP_001294039.1 | 2 - 3 | 253 | 61 | ||
| NM_069202.6 | NP_501603.3 | 14 - 15 | 5503 | 61 | ||
| NM_182275.7 | NP_872075.2 | 13 - 14 | 5022 | 61 | ||
More Information
Set Membership: |
|

wnk-1
Genomic Map