Assay Details
| Gene Symbol: pes-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028235.3 | NP_001023406.1 | 2 - 3 | 182 | 55 | |
| NM_001383089.1 | 2 - 3 | 185 | 55 | |||
| GenBank mRNA | Z28376.1 | - | 81 | 55 | ||
| Z28377.1 | - | 178 | 55 | |||
More Information
Set Membership: |
|

pes-1
Genomic Map