Assay Details
| Gene Symbol: flp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069191.7 | NP_501592.1 | 5 - 6 | 475 | 98 | |
| NM_182277.8 | NP_872077.1 | 5 - 6 | 429 | 98 | ||
| NM_182278.7 | NP_872078.1 | 5 - 6 | 355 | 98 | ||
More Information
Set Membership: |
|

flp-1
Genomic Map