Assay Details
| Gene Symbol: flp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182277.8 | NP_872077.1 | 3 - 4 | 184 | ||
| NM_182278.7 | NP_872078.1 | 3 - 4 | 247 | 118 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-1| Gene Symbol: flp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182277.8 | NP_872077.1 | 3 - 4 | 184 | ||
| NM_182278.7 | NP_872078.1 | 3 - 4 | 247 | 118 | ||
Set Membership: |
|