Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268365.3 | NP_001255294.1 | 26 - 27 | 4325 | 133 | |
| NM_001268366.3 | NP_001255295.1 | 21 - 22 | 3531 | 133 | ||
| NM_001268369.2 | NP_001255298.1 | 22 - 23 | 3519 | 133 | ||
| NM_001307278.3 | NP_001294207.1 | 22 - 23 | 3729 | 133 | ||
| NM_001372869.1 | NP_001359805.1 | 22 - 23 | 3519 | 133 | ||
| NM_001380324.1 | NP_001367366.1 | 23 - 24 | 3711 | 133 | ||
| NM_001380325.1 | NP_001367367.1 | 25 - 26 | 3952 | 133 | ||
| NM_001380326.1 | NP_001367365.1 | 3 - 4 | 313 | 133 | ||
| NM_068785.7 | NP_501186.5 | 22 - 23 | 3678 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380326.1 transcripts.
Set Membership: |
|

unc-82
Genomic Map