Assay Details
| Gene Symbol: unc-82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268369.2 | NP_001255298.1 | 26 - 27 | 4095 | 107 | |
| NM_001380326.1 | NP_001367365.1 | 7 - 8 | 934 | 107 | ||
More Information
Set Membership: |
|

unc-82
Genomic Map