Assay Details
| Gene Symbol: npr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372906.2 | NP_001359554.1 | 7 - 8 | 1063 | 91 | |
| NM_001380412.1 | NP_001368470.1 | 7 - 8 | 1051 | 91 | ||
| NM_069300.5 | NP_501701.2 | 9 - 10 | 1243 | 91 | ||
More Information
Set Membership: |
|

npr-2
Genomic Map