Assay Details
| Gene Symbol: ntl-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307177.4 | NP_001294106.1 | 5 - 6 | 1175 | 113 | |
| NM_001380548.1 | NP_001367046.1 | 2 - 3 | 230 | 113 | ||
| NM_070459.5 | NP_502860.2 | 5 - 6 | 1169 | 113 | ||
More Information
Set Membership: |
|

ntl-11
Genomic Map