Assay Details
Gene Symbol: ntl-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307177.4 | NP_001294106.1 | 6 - 7 | 1380 | 95 | |
NM_001307178.3 | NP_001294107.1 | 1 - 2 | 186 | 95 | ||
NM_001380548.1 | NP_001367046.1 | 3 - 4 | 435 | 95 | ||
NM_070459.5 | NP_502860.2 | 6 - 7 | 1374 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
