Assay Details
Gene Symbol: ntl-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307177.4 | NP_001294106.1 | 7 - 8 | 1605 | 87 | |
NM_001307178.3 | NP_001294107.1 | 2 - 3 | 411 | 87 | ||
NM_001380548.1 | NP_001367046.1 | 4 - 5 | 660 | 87 | ||
NM_070459.5 | NP_502860.2 | 7 - 8 | 1599 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
