Assay Details
| Gene Symbol: eef-1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383394.1 | 3 - 4 | 521 | 66 | ||
| NM_073399.6 | NP_505800.1 | 3 - 4 | 598 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eef-1G| Gene Symbol: eef-1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383394.1 | 3 - 4 | 521 | 66 | ||
| NM_073399.6 | NP_505800.1 | 3 - 4 | 598 | 66 | ||
Set Membership: |
|