Assay Details
| Gene Symbol: eef-1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047635.6 | NP_001041100.1 | 4 - 5 | 548 | 58 | |
| NM_001383394.1 | 4 - 5 | 661 | 58 | |||
| NM_073399.6 | NP_505800.1 | 4 - 5 | 738 | 58 | ||
More Information
Set Membership: |
|

eef-1G
Genomic Map