Assay Details
| Gene Symbol: rol-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269168.3 | NP_001256097.1 | 4 - 5 | 818 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rol-3| Gene Symbol: rol-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269168.3 | NP_001256097.1 | 4 - 5 | 818 | 110 | |
Set Membership: |
|