Assay Details
| Gene Symbol: slc-25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307806.4 | NP_001294735.1 | 1 - 2 | 208 | 65 | |
| NM_001373160.1 | NP_001359577.1 | 1 - 2 | 220 | 65 | ||
| NM_182334.4 | NP_872134.1 | 1 - 2 | 221 | 65 | ||
More Information
Set Membership: |
|

slc-25A29
Genomic Map