Assay Details
| Gene Symbol: slc-25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307806.4 | NP_001294735.1 | 3 - 4 | 627 | 88 | |
| NM_001373160.1 | NP_001359577.1 | 3 - 4 | 639 | 88 | ||
| NM_182334.4 | NP_872134.1 | 3 - 4 | 640 | 88 | ||
More Information
Set Membership: |
|

slc-25A29
Genomic Map