Assay Details
| Gene Symbol: slc-25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182334.4 | NP_872134.1 | 5 - 6 | 1071 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc-25A29| Gene Symbol: slc-25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182334.4 | NP_872134.1 | 5 - 6 | 1071 | 140 | |
Set Membership: |
|