Assay Details
| Gene Symbol: nfs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060411.7 | NP_492812.2 | 9 - 10 | 1171 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nfs-1| Gene Symbol: nfs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060411.7 | NP_492812.2 | 9 - 10 | 1171 | 83 | |
Set Membership: |
|