Assay Details
| Gene Symbol: F49D11.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060446.2 | NP_492847.1 | 2 - 3 | 313 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F49D11.2| Gene Symbol: F49D11.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060446.2 | NP_492847.1 | 2 - 3 | 313 | 125 | |
Set Membership: |
|