Assay Details
| Gene Symbol: spe-38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060452.8 | NP_492853.2 | 1 - 2 | 164 | 77 | |
| Gene Symbol: impt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026611.4 | NP_001021782.1 | 1 - 2 | 168 | 77 | |
| NM_001026612.4 | NP_001021783.1 | 1 - 2 | 169 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_060452.8 transcripts.
Set Membership: |
|

spe-38
Genomic Map