Assay Details
| Gene Symbol: sri-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026253.3 | NP_001021424.1 | 6 - 7 | 855 | 99 | |
| NM_001026254.4 | NP_001021425.1 | 4 - 5 | 775 | 99 | ||
More Information
Set Membership: |
|

sri-5
Genomic Map