Assay Details
| Gene Symbol: cdk-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060505.4 | NP_492906.2 | 7 - 8 | 1109 | 120 | |
| NM_060506.5 | NP_492907.2 | 7 - 8 | 1109 | 120 | ||
More Information
Set Membership: |
|

cdk-9
Genomic Map