Assay Details
| Gene Symbol: cdk-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060505.4 | NP_492906.2 | 8 - 9 | 1152 | 92 | |
| NM_060506.5 | NP_492907.2 | 8 - 9 | 1152 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdk-9| Gene Symbol: cdk-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060505.4 | NP_492906.2 | 8 - 9 | 1152 | 92 | |
| NM_060506.5 | NP_492907.2 | 8 - 9 | 1152 | 92 | ||
Set Membership: |
|