Assay Details
| Gene Symbol: wbp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060511.4 | NP_492912.1 | 5 - 6 | 1357 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wbp-4| Gene Symbol: wbp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060511.4 | NP_492912.1 | 5 - 6 | 1357 | 96 | |
Set Membership: |
|