Assay Details
| Gene Symbol: aat-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026617.4 | NP_001021788.1 | 3 - 4 | 378 | 72 | |
| NM_001026618.5 | NP_001021789.1 | 3 - 4 | 380 | 72 | ||
More Information
Set Membership: |
|

aat-9
Genomic Map