Assay Details
| Gene Symbol: aat-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026617.4 | NP_001021788.1 | 4 - 5 | 455 | 97 | |
| NM_001026618.5 | NP_001021789.1 | 4 - 5 | 457 | 97 | ||
More Information
Set Membership: |
|

aat-9
Genomic Map