Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 12 - 13 | 2941 | 122 | |
| NM_001306311.3 | NP_001293240.1 | 1 - 2 | 142 | 122 | ||
| NM_001306313.4 | NP_001293242.1 | 16 - 17 | 3619 | 122 | ||
| NM_001306314.3 | NP_001293243.1 | 12 - 13 | 2941 | 122 | ||
| NM_001383731.1 | 12 - 13 | 2992 | 122 | |||
| NM_060545.6 | NP_492946.1 | 16 - 17 | 3681 | 122 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map