Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 18 - 19 | 4473 | 60 | |
| NM_001306311.3 | NP_001293240.1 | 7 - 8 | 1674 | 60 | ||
| NM_001306313.4 | NP_001293242.1 | 21 - 22 | 5118 | 60 | ||
| NM_001306314.3 | NP_001293243.1 | 17 - 18 | 4440 | 60 | ||
| NM_001383731.1 | 17 - 18 | 4491 | 60 | |||
| NM_060545.6 | NP_492946.1 | 22 - 23 | 5213 | 60 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map