Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 22 - 23 | 5472 | 73 | |
| NM_001306311.3 | NP_001293240.1 | 11 - 12 | 2673 | 73 | ||
| NM_001306313.4 | NP_001293242.1 | 25 - 26 | 6117 | 73 | ||
| NM_001306314.3 | NP_001293243.1 | 21 - 22 | 5439 | 73 | ||
| NM_001383731.1 | 21 - 22 | 5490 | 73 | |||
| NM_060545.6 | NP_492946.1 | 26 - 27 | 6212 | 73 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map