Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 30 - 31 | 7973 | 64 | |
| NM_001306311.3 | NP_001293240.1 | 19 - 20 | 5174 | 64 | ||
| NM_001306312.3 | NP_001293241.1 | 7 - 8 | 2360 | 64 | ||
| NM_001306313.4 | NP_001293242.1 | 33 - 34 | 8618 | 64 | ||
| NM_001306314.3 | NP_001293243.1 | 29 - 30 | 7940 | 64 | ||
| NM_001383731.1 | 29 - 30 | 7991 | 64 | |||
| NM_060545.6 | NP_492946.1 | 34 - 35 | 8713 | 64 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map