Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 35 - 36 | 9031 | 103 | |
| NM_001306311.3 | NP_001293240.1 | 24 - 25 | 6232 | 103 | ||
| NM_001306312.3 | NP_001293241.1 | 12 - 13 | 3418 | 103 | ||
| NM_001306313.4 | NP_001293242.1 | 38 - 39 | 9676 | 103 | ||
| NM_001306314.3 | NP_001293243.1 | 34 - 35 | 8998 | 103 | ||
| NM_001383731.1 | 34 - 35 | 9049 | 103 | |||
| NM_060545.6 | NP_492946.1 | 39 - 40 | 9771 | 103 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map