Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 37 - 38 | 9233 | 65 | |
| NM_001306311.3 | NP_001293240.1 | 26 - 27 | 6434 | 65 | ||
| NM_001306312.3 | NP_001293241.1 | 14 - 15 | 3620 | 65 | ||
| NM_001306313.4 | NP_001293242.1 | 40 - 41 | 9878 | 65 | ||
| NM_001306314.3 | NP_001293243.1 | 36 - 37 | 9200 | 65 | ||
| NM_001383731.1 | 36 - 37 | 9251 | 65 | |||
| NM_060545.6 | NP_492946.1 | 41 - 42 | 9973 | 65 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map